Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | SNAP23 |
|---|---|
| Concentration | 0.57 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6630
![]()
|
Proteintech Group Inc
108251AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Synaptosomal-associated proteins (SNAPs) are cytosolic proteins that play a key role in in the process of membrane fusion in intracellular vesicle trafficking. In eukaryotic cells, the SNARE (soluble N-ethylmaleimide-sensitive factor attachment protein receptor) complex is critical to membrane docking and fusion and is believed to impart some degree of specificity between vesicle SNARE (v-SNARE) and target organelle SNARE (t-SNARE). In neurons and neuroendocrine cells, the SNARE complex consists of the integral membrane proteins VAMP (vesicle-associated membrane protein), syntaxin and SNAP-25. In non-neuronal tissue, a SNAP-25 homolog, SNAP-23, functionally replaces SNAP-25 in the SNARE complex. Studies show that VAMP, syntaxin and SNAP-23 are required for SNARE function and that this complex exists as a heterotrimer of the three proteins. In insulin signaling pathways, studies suggest that the translocation of GLUT4 in adipocytes requires that functional SNAP-23 be present in SNARE complexes.Specifications
| SNAP23 | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O00161, O09044, O70377 | |
| 20619, 64630, 8773 | |
| SNAP23 Fusion Protein Ag1269 | |
| Primary | |
| -20°C | |
| SNAP23 |
| 0.57 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| HsT17016, SNAP 23, SNAP23, SNAP23A, SNAP23B | |
| Snap23 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title