Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP25 Antibody (4E11), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N104344APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N104344APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N104344APC7 Supplier Novus Biologicals Supplier No. NBP104344APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNAP25 Monoclonal specifically detects SNAP25 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SNAP25
Applications Flow Cytometry
Classification Monoclonal
Clone 400000000000
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa
Gene Symbols SNAP25
Host Species Mouse
Immunogen Recombinant human SNAP25 (1-206aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6616.0
Test Specificity Anti-human SNAP25 mAb, clone 4E11, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human SNAP25 protein.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.