Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP25 Antibody (SP12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600692
Description
SNAP25 Monoclonal specifically detects SNAP25 in Human, Rat, Porcine, Hamster, Gerbil samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SNAP25 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:5000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:2000-1:5000 | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| SNAP25 (SP12) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SP12 | |
| Unconjugated | |
| P60880 | |
| SNAP25 | |
| Crude synaptic immunoprecipitate (Human). | |
| 0.1 mg | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 6616 | |
| Human, Rat, Pig, Hamster, Gerbil | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction