Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP25 Antibody (SP12) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen SNAP25
Clone SP12
Concentration 1.0 mg/mL
Dilution Western Blot 1:500-1:5000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:2000-1:5000
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018250
SDP
View Documents
Novus Biologicals
NB6006920.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB600692
SDP
View Documents
Novus Biologicals
NB600692
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

SNAP25 Monoclonal specifically detects SNAP25 in Human, Rat, Porcine, Hamster, Gerbil samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SNAP25
1.0 mg/mL
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
P60880
6616
Crude synaptic immunoprecipitate (Human).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SP12
Western Blot 1:500-1:5000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:2000-1:5000
Monoclonal
Purified
RUO
Rat, Gerbil, Hamster, Human, Pig
bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa
SNAP25
IgG1
Protein A purified
SNAP25 (SP12)
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.