Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP25 Antibody (SP12), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB015063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015063 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015063 Supplier Novus Biologicals Supplier No. NBP281057JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SNAP25 Monoclonal specifically detects SNAP25 in Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SNAP25
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SP12
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa
Gene Symbols SNAP25
Host Species Rabbit
Immunogen Recombinant monoclonal antibody to SNAP25.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6616
Test Specificity This antibody binds to native and denatured SNAP25.
Target Species Human, Mouse, Rat, Pig, Bovine, Feline, Guinea Pig, Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.