Learn More
Description
Specifications
Specifications
| Antigen | SNAP45 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Proximal sequence element-binding transcription factor subunit delta, PSE-binding factor subunit delta, PTF subunit delta, PTFdelta, small nuclear RNA activating complex, polypeptide 2, 45kD, small nuclear RNA activating complex, polypeptide 2, 45kDa, Small nuclear RNA-activating complex polypeptide 2, SNAP45snRNA-activating protein complex 45 kDa subunit, SNAPc 45 kDa subunit, SNAPc subunit 2, snRNA-activating protein complex subunit 2 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human SNAP45 (NP_003074). Peptide sequence STEEDFAVDFEKIYKYLSSVSRSGRSPELSAAESAVVLDLLMSLPEELPL |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
