Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNF1LK2/SIK2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056458 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056458 0.1 mg
NB10056458T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056458 Supplier Novus Biologicals Supplier No. NB10056458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNF1LK2/SIK2 Polyclonal specifically detects SNF1LK2/SIK2 in Bovine, Canine, Human, Mouse, Primate, Rat samples. It is validated for Western Blot.

Specifications

Antigen SNF1LK2/SIK2
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9H0K1
Gene Alias DKFZp434K1115, EC 2.7.11, EC 2.7.11.1, KIAA0781SIK-2, LOH11CR1I, QIKSNF1LK2, Qin-induced kinase, salt-inducible kinase 2, Salt-inducible protein kinase 2, salt-inducible serine/threonine kinase 2, serine/threonine-protein kinase SIK2, Serine/threonine-protein kinase SNF1-like kinase 2, SNF1-like kinase 2
Gene Symbols SIK2
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human SIK2 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23235
Test Specificity Human SIK2 is a 926 amino acid protein. The approximately 120 kD band is higher than the theoretical expected size, but this band, as well as the approximately 25 kD lower band, are blocked by immunizing peptide.
Target Species Bovine, Mouse, Canine, Human, Mouse, Primate, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.