Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNF1LK2/SIK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $426.00
Specifications
Antigen | SNF1LK2/SIK2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056458T
![]() |
Novus Biologicals
NB10056458SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056458
![]() |
Novus Biologicals
NB10056458 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
SNF1LK2/SIK2 Polyclonal specifically detects SNF1LK2/SIK2 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.Specifications
SNF1LK2/SIK2 | |
Western Blot 1-3 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Canine, Primate | |
Q9H0K1 | |
23235 | |
A portion of amino acids 100-150 of human SIK2 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS with 0.05% Sodium Azide | |
DKFZp434K1115, EC 2.7.11, EC 2.7.11.1, KIAA0781SIK-2, LOH11CR1I, QIKSNF1LK2, Qin-induced kinase, salt-inducible kinase 2, Salt-inducible protein kinase 2, salt-inducible serine/threonine kinase 2, serine/threonine-protein kinase SIK2, Serine/threonine-protein kinase SNF1-like kinase 2, SNF1-like kinase 2 | |
SIK2 | |
IgG | |
Protein G purified | |
Human SIK2 is a 926 amino acid protein. The approximately 120 kD band is higher than the theoretical expected size, but this band, as well as the approximately 25 kD lower band, are blocked by immunizing peptide. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title