Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNHG28 Antibody, Novus Biologicals™
SDP

Catalog No. NBP276544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276544 100 μL
NB402744 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP276544 Supplier Novus Biologicals Supplier No. NBP276544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNHG28 Polyclonal specifically detects SNHG28 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SNHG28
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias C1orf204, Small Nucleolar RNA Host Gene 28, VSIG8-OT1
Gene Symbols SNHG28
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: SGVDFPQVSAWMRALPSPDCPGLRTTGEQMQKLLLKENKVKTRKSKRRSGEGSHLTTSILEQ
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 284677.0
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.