Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SNOG

Click to view available options
Quantity:
10 mg
50 mg
Description
A carrier of NO, relaxing smooth muscle and inhibiting platelet aggregation.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 336.32 |
| Purity | >98% |
| Molecular Weight (g/mol) | 336.319 |
| Chemical Name or Material | SNOG |
Safety and Handling
| Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction