Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRNP25 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB327504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327504 Supplier Novus Biologicals Supplier No. NBP306000MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNRNP25 Polyclonal antibody specifically detects SNRNP25 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SNRNP25
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C16orf33, chromosome 16 open reading frame 33, FLJ22940, Minus-99 protein, small nuclear ribonucleoprotein 25kDa (U11/U12), U11/U12 small nuclear ribonucleoprotein 25 kDa protein, U11/U12 snRNP 25 kDa protein, U11/U12 snRNP 25K, U11/U12-25K
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SNRNP25 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79622
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.