Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRNP40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120664
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5120664 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5120664

Supplier: Invitrogen™ PA5120664

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, HeLa, Mouse brain, Mouse testis. Immunogen sequence: MIEQQKRKGP ELPLVPVKRQ RHELLLGAGS GPGAGQQQAT PGALLQAGPP RCSSLQAPIM LLSGHEGEVY CCKFHPNGST LASAGFDRLI LLWNVYGDCD NYATLKGHSG AVMELHYNTD GSMLFSASTD KTVAVWDSET GERVKRLKGH TSFVNSCYPA RRGPQLVCTG SDDGTVKLWD IRKKAAIQTF QNTYQVLAVT FNDTSDQIIS GGIDNDIKVW DLRQNKLTYT MRGHADSVTG LSLSSEGSYL LSNAMDNTVR VWDVRPFAPK ERCVKIFQGN VHNFEKNLLR CSWSPDGSKI AAGSADRFVY VWDTTSRRIL YKLPGHAGSI NEVAFHPDEP IIISASSDKR LYMGEIQ.

This gene encodes a component of the U5 small nuclear ribonucleoprotein (snRNP) particle. The U5 snRNP is part of the spliceosome, a multiprotein complex that catalyzes the removal of introns from pre-messenger RNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP40
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNRNP40
Gene Accession No. Q6PE01, Q96DI7
Gene Alias 0610009C03Rik; 38 kDa-splicing factor; 40K; HPRP8BP; Prp8-binding protein; Prp8bp; PRPF8BP; RGD1309198; RP11-490K7.3; SFP38; small nuclear ribonucleoprotein 40 (U5); small nuclear ribonucleoprotein 40kDa (U5); small nuclear ribonucleoprotein U5 subunit 40; small nuclear ribonucleoprotein, U5 40kDa subunit; Snrnp40; SPF38; U5 small nuclear ribonucleoprotein 40 kDa protein; U5 snRNP 40 kDa protein; U5 snRNP-specific 40 kDa protein; U5 snRNP-specific 40 kDa protein (hPrp8-binding); U5 snRNP-specific protein (Prp8-binding); U5-40K; U5-40kD protein; WD repeat domain 57 (U5 snRNP specific); WD repeat-containing protein 57; Wdr57
Gene Symbols SNRNP40
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-357 of human SNRNP40 (NP_004805.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66585, 9410
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.