Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRP70 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237896
Description
SNRP70 Polyclonal specifically detects SNRP70 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SNRP70 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
P08621 | |
SNRNP70 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RDPIPYLPPLEKLPHEKHHNQPYCGIAPYIREFEDPRDAPPPTRAETREERMERKRREKIERRQQEVETELKMWDPHNDPNAQ | |
0.1 mL | |
Primary | |
Specificity of human SNRP70 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RNPU1ZU170K, RPU1U1AP, small nuclear ribonucleoprotein 70kDa (U1), Snp1, snRNP70, SNRP70U1 small nuclear ribonucleoprotein 70 kDa, U1 snRNP 70 kDa, U1-70Ksmall nuclear ribonucleoprotein 70kDa (RNP antigen), U1AP1, U1RNP | |
Rabbit | |
Affinity Purified | |
RUO | |
6625 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction