Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRP70 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SNRP70 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SNRP70 Polyclonal specifically detects SNRP70 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SNRP70 | |
Polyclonal | |
Rabbit | |
Human | |
P08621 | |
6625 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RDPIPYLPPLEKLPHEKHHNQPYCGIAPYIREFEDPRDAPPPTRAETREERMERKRREKIERRQQEVETELKMWDPHNDPNAQ | |
Primary | |
Specificity of human SNRP70 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RNPU1ZU170K, RPU1U1AP, small nuclear ribonucleoprotein 70kDa (U1), Snp1, snRNP70, SNRP70U1 small nuclear ribonucleoprotein 70 kDa, U1 snRNP 70 kDa, U1-70Ksmall nuclear ribonucleoprotein 70kDa (RNP antigen), U1AP1, U1RNP | |
SNRNP70 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title