Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRPA1 Antibody (SR1474), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32243250UL
Description
SNRPA1 Monoclonal antibody specifically detects SNRPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SNRPA1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1474 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Lea1, small nuclear ribonucleoprotein polypeptide A', U2 small nuclear ribonucleoprotein A', U2 small nuclear ribonucleoprotein polypeptide A', U2 snRNP A' | |
A synthesized peptide derived from human SNRPA1 (Uniprot #: P09661) | |
50 μg | |
Core ESC Like Genes, Stem Cell Markers | |
6627 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction