Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRPA1 Antibody (SR1474), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50
Specifications
Antigen | SNRPA1 |
---|---|
Clone | SR1474 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
SNRPA1 Monoclonal antibody specifically detects SNRPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SNRPA1 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Lea1, small nuclear ribonucleoprotein polypeptide A', U2 small nuclear ribonucleoprotein A', U2 small nuclear ribonucleoprotein polypeptide A', U2 snRNP A' | |
A synthesized peptide derived from human SNRPA1 (Uniprot #: P09661) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1474 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6627 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title