Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPB2 Antibody (OTI2A5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274260A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274260A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274260A94 Supplier Novus Biologicals Supplier No. NBP274260AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNRPB2 Monoclonal specifically detects SNRPB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SNRPB2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2A5
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC24807, MGC45309, Msl1, small nuclear ribonucleoprotein polypeptide B, small nuclear ribonucleoprotein polypeptide B′′, small nuclear ribonucleoprotein polypeptide B2, U2 small nuclear ribonucleoprotein B′′, U2 snRNP B′′
Gene Symbols SNRPB2
Host Species Mouse
Immunogen Full length recombinant protein of human SNRPB2 (NP_937863) produced in E.coli.
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6629
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.