Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119422
This item is not returnable.
View return policy
Description
SNRPD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.
Specifications
SNRPD1 | |
Polyclonal | |
Unconjugated | |
SNRPD1 | |
AA407109; AL023031; CHUNP6882; fk26a01; HsT2456; small nuclear ribonucleoprotein D1; small nuclear ribonucleoprotein D1 polypeptide; small nuclear ribonucleoprotein D1 polypeptide 16kDa; small nuclear ribonucleoprotein D1 polypeptide 16kDa pseudogene; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; small nuclear ribonucleoprotein Sm D1; Sm-D autoantigen; SMD1; sm-D1; snprd1; snRNP core protein D1; snrnpd1; SNRPD; Snrpd1; snrpd1 protein; wu:fk26a01; zgc:86929 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100524872, 291794, 6632 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P62314 | |
SNRPD1 | |
Recombinant protein Small Nuclear Ribonucleoprotein Polypeptide D1. The antigen corresponds to amino acid range 1-119 of the target protein. | |
200 μL | |
Primary | |
Human, Rat, Pig | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction