Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNRPD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | SNRPD1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119422
|
Invitrogen™
PA5119422 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121593
|
Invitrogen™
PA5121593 |
20 μL |
Each for $178.40
|
|
|||||
Description
SNRPD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.Specifications
SNRPD1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Rat, Pig | |
SNRPD1 | |
AA407109; AL023031; CHUNP6882; fk26a01; HsT2456; small nuclear ribonucleoprotein D1; small nuclear ribonucleoprotein D1 polypeptide; small nuclear ribonucleoprotein D1 polypeptide 16kDa; small nuclear ribonucleoprotein D1 polypeptide 16kDa pseudogene; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; small nuclear ribonucleoprotein Sm D1; Sm-D autoantigen; SMD1; sm-D1; snprd1; snRNP core protein D1; snrnpd1; SNRPD; Snrpd1; snrpd1 protein; wu:fk26a01; zgc:86929 | |
SNRPD1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P62314 | |
100524872, 291794, 6632 | |
Recombinant protein Small Nuclear Ribonucleoprotein Polypeptide D1. The antigen corresponds to amino acid range 1-119 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title