Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPN Antibody, Novus Biologicals™
SDP

Catalog No. NBP220439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220439 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220439 Supplier Novus Biologicals Supplier No. NBP220439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNRPN Polyclonal specifically detects SNRPN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SNRPN
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P63162
Gene Alias DKFZp686C0927, DKFZp686M12165, DKFZp761I1912, DKFZp762N022, FLJ39265, HCERN3FLJ33569, MGC29886, Prader-Willi syndrome chromosome region, PWCR, RT-LI, Sm protein D, Sm protein N, small nuclear ribonucleoprotein polypeptide N, small nuclear ribonucleoprotein-associated protein N, Sm-D, sm-N, SmN, SMNFLJ36996, SNRNP-N, SNURF-SNRPN, tissue-specific splicing protein, Tissue-specific-splicing protein
Gene Symbols SNRPN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SNRPN. The exact sequence is proprietary.
Molecular Weight of Antigen 25 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 6638
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Pig (100%), Rat (100%), Bovine (100%), Chimpanzee (100%), Rhesus Monkey (100%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.