Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SNS 032
GREENER_CHOICE

Catalog No. p-4817039
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk2, cdk7 and cdk9

Cyclin dependent kinase (cdk) inhibitor (reported IC50 values are 0.004 - 0.048, 0.0045 - 0.05, 0.062 - 0.29, 0.2-0.48, 0.4 and 0.45 μM for cdk9, cdk2, cdk7, cdk1, cdk5 and cdk4, respectively). Displays no activity against 190 additional kinases (IC50 >1 μM). Arrests the cell cycle at G2/M; inhibits transcription, proliferation and colony formation, and induces apoptosis in RPMI-8226 multiple myeloma cells. Prevents tumor cell-induced VEGF secretion and in vitro angiogenesis.

Chemical Identifiers

CAS 345627-80-7
Molecular Formula C17H24N4O2S2

Specifications

Quantity 10 mg
Formula Weight 380.53
Purity >99%
Molecular Weight (g/mol) 380.525
Chemical Name or Material SNS 032
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.