Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNTF (cleaved spectrin) Rabbit anti-Mouse, Porcine, Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$193.44 - $456.56
Specifications
Antigen | SNTF (cleaved spectrin) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN2264-10UL
|
MilliporeSigma
ABN226410UL |
10 μL |
Each for $193.44
|
|
|||||
ABN2264-50UL
|
MilliporeSigma
ABN226450UL |
50 μL |
Each for $456.56
|
|
|||||
Description
Anti-SNTF (cleaved spectrin), Cat. No. ABN2264, is a highly specific rabbit polyclonal antibody that targets cleaved Spectrin alpha chain, non-erythrocytic 1 and has been tested for use in ELISA, Immunohistochemistry, and Western Blotting.Specifications
SNTF (cleaved spectrin) | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
Rabbit polyclonal antiserum with 0.05% sodium azide. | |
NP_003118 | |
KLH-conjugated linear peptide corresponding to calpain-generated neoepitope in SNTF (CAQQEVY). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Serum | |
RUO | |
Mouse, Pig, Human | |
Spectrin alpha chain non-erythrocytic 1;Alpha-II spectrin;Fodrin alpha chain;Spectrin non-erythroid alpha subunit;Calpain-derived alpha-II spectrin N-terminal fragment | |
SPTAN1;NEAS;SPTA2 | |
IgG | |
Unpurified | |
This rabbit polyclonal antibody specifically detects calpain-generated neoepitope in SNTF. It targets a neoepitope of 6 amino acids at the carboxyl-end of the stable calpain-derived alpha-spectrin. | |
∽160 kDa observed. Calculated molecular weight of full length protein is 284.54 kDa. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title