Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNTG1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1741920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1741920 20 μL
NBP174119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1741920 Supplier Novus Biologicals Supplier No. NBP17411920UL

Rabbit Polyclonal Antibody

SNTG1 Polyclonal specifically detects SNTG1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen SNTG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q925E1
Gene Alias G1SYNsyntrophin-4, gamma-1-syntrophin, gamma1-syntrophin, SYN4syntrophin 4, syntrophin, gamma 1, Syntrophin-4
Gene Symbols SNTG1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the N terminal of Sntg1. Immunizing peptide sequence KLPVNEDCACAPSDQSSGTSSPLCDSGLHLNYHPNNTDTLSCSSWPTSPG.
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54212
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.