Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNW1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002368 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002368 Supplier Novus Biologicals Supplier No. NB1002368
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNW1 Polyclonal specifically detects SNW1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SNW1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q13573
Gene Alias Bx42, homolog of Drosophila BX42, MGC119379, NCOA-62, Nuclear protein SkiP, Nuclear receptor coactivator NCoA-62, nuclear receptor coactivator, 62-kD, Prp45, PRPF45, SKI interacting protein, Ski-interacting protein, SKIPSKIIP, SNW domain containing 1, SNW domain-containing protein 1
Gene Symbols SNW1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and the C-terminus (residue 536) of human Nuclear Receptor Coactivator, 62 kD (SKIIP, SKI Interacting Protein) using the numbering given in entry NP_036377.1 (GeneID 22938).
Molecular Weight of Antigen 61 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22938
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.