Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX16 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP261226 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261226 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261226 Supplier Novus Biologicals Supplier No. NBP261226

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

SNX16 Polyclonal specifically detects SNX16 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen SNX16
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation 8 - 15 ul/mg lysate
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias DKFZp666H147, sorting nexin 16, sorting nexin-16
Gene Symbols SNX16
Host Species Rabbit
Immunogen 100% Sequence Identity Orangutan between 294 and 344
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64089
Target Species Human
Content And Storage Store the unopened product at 2 - 8degrees C. Protect from light. Do not use past expiration date.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.