Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX18 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220443 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220443 Supplier Novus Biologicals Supplier No. NBP220443
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNX18 Polyclonal specifically detects SNX18 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SNX18
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias FLJ11997, FLJ61062, MGC150827, MGC150829, SH3 and PX domain-containing protein 3B, SH3PX2, SH3PXD3BFLJ32560, SNAG1, sorting nexin 18, sorting nexin associated golgi protein 1, sorting nexin-18, Sorting nexin-associated Golgi protein 1
Gene Symbols SNX18
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SNX18. The exact sequence is proprietary.
Molecular Weight of Antigen 69 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112574
Test Specificity Human, Mouse, Primate. Species reactivity based on sequence identity: Rat (100%), Rhesus Monkey (100%).
Target Species Human, Mouse, Rat, Monkey
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.