Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX18 Antibody, Novus Biologicals™
SDP

Catalog No. NBP182361 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182361 0.1 mL
NB433432 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP182361 Supplier Novus Biologicals Supplier No. NBP182361
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNX18 Polyclonal specifically detects SNX18 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen SNX18
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ11997, FLJ61062, MGC150827, MGC150829, SH3 and PX domain-containing protein 3B, SH3PX2, SH3PXD3BFLJ32560, SNAG1, sorting nexin 18, sorting nexin associated golgi protein 1, sorting nexin-18, Sorting nexin-associated Golgi protein 1
Gene Symbols SNX18
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:DDEWDDSSTVADEPGALGSGAYPDLDGSSSAGVGAAGRYRLSTRSDLSLGSRGGSVPPQHHPSGPKSSATVSRNLN
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112574
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.