Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX19 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120976 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120976 Supplier Novus Biologicals Supplier No. NBP120976
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SNX19 Polyclonal specifically detects SNX19 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen SNX19
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_055573.2
Gene Alias DKFZp667I205, KIAA0254CHET8, sorting nexin 19, sorting nexin-19
Gene Symbols SNX19
Host Species Goat
Immunogen Peptide with sequence C-TTSASDTPGNSKR corresponding to internal region according to NP_055573.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 399979
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.