Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX27 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP145282C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145282C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145282C Supplier Novus Biologicals Supplier No. NBP145282C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNX27 Polyclonal specifically detects SNX27 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SNX27
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q96L92
Gene Alias KIAA0488MGC126873, methamphetamine-responsive transcript 1, MGC126871, MGC20471, MRT1, MY014, sorting nexin family member 27, sorting nexin-27
Gene Symbols SNX27
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SNX27 (within residues 150-200). [Swiss-Prot# Q96L92]
Molecular Weight of Antigen 64 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 81609
Test Specificity Human and mouse. Immunogen sequence has 94% identity to rat and bovine.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.