Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB10056637
Description
SOCS-1 Polyclonal specifically detects SOCS-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
| SOCS-1 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide | |
| JABTec-interacting protein 3, JAK binding protein, JAK-binding protein, SOCS-1CIS1, SSI-1CISH1, SSI1cytokine-inducible SH2 protein 1, STAT induced SH3 protein 1, STAT-induced STAT inhibitor 1, suppressor of cytokine signaling 1, TIP-3, TIP3Cish1 | |
| Rabbit | |
| 23.55 kDa | |
| 0.2 mL | |
| Primary | |
| This should cross-react with rat SOCS1, based on amino acid sequence. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1:500-1:1000 | |
| O15524 | |
| SOCS1 | |
| A synthetic peptide corresponding to amino acids 54-68 (HFRTFRSHADYRRIT) of human SOCS1 was used as immunogen (NP_003736.1) | |
| Unpurified | |
| RUO | |
| 8651 | |
| Human, Mouse | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction