Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$149.33 - $480.74

Specifications

Antigen SOCS-1
Concentration 0.5 mg/mL
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056637T
SDP
View Documents
Novus Biologicals
NB10056637SS
0.05 mL
Each for $149.33
Only null left
Add to Cart
 
NB10056637
SDP
View Documents
Novus Biologicals
NB10056637
0.2 mL
Each for $480.74
Only null left
Add to Cart
 
Description

Description

SOCS-1 Polyclonal specifically detects SOCS-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

SOCS-1
Western Blot 1:500-1:1000
Polyclonal
Antisera
RUO
Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide
JABTec-interacting protein 3, JAK binding protein, JAK-binding protein, SOCS-1CIS1, SSI-1CISH1, SSI1cytokine-inducible SH2 protein 1, STAT induced SH3 protein 1, STAT-induced STAT inhibitor 1, suppressor of cytokine signaling 1, TIP-3, TIP3Cish1
SOCS1
IgG
Unpurified
This should cross-react with rat SOCS1, based on amino acid sequence.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human, Mouse
O15524
8651
A synthetic peptide corresponding to amino acids 54-68 (HFRTFRSHADYRRIT) of human SOCS1 was used as immunogen (NP_003736.1)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
23.55 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.