Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS-3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227210 0.1 mg
NBP227210SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227210 Supplier Novus Biologicals Supplier No. NBP227210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOCS-3 Polyclonal specifically detects SOCS-3 in Human, Feline, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SOCS-3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-5ug/ml, Immunohistochemistry-Paraffin 5ug/ml∼
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3
Gene Symbols SOCS3
Host Species Rabbit
Immunogen This antibody was raised against SOCS3
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9021
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, rhesus monkey, orangutan, european pole-cat, and chimpanzee, 94% in new world monkey, and pig, 89% in panda, dog, Chinese hamster, and horse, 83% in cow, african bush elephant, mouse, sheep, and rat, and 61% homologous in guinea pig.
Target Species Human, Feline, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.