Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS-7/Nck/NAP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15890220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15890220 20 μL
NBP158902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15890220 Supplier Novus Biologicals Supplier No. NBP15890220UL

Rabbit Polyclonal Antibody

SOCS-7/Nck/NAP4 Polyclonal specifically detects SOCS-7/Nck/NAP4 in Human samples. It is validated for Western Blot.

Specifications

Antigen SOCS-7/Nck/NAP4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O14512
Gene Alias NAP-4, NAP4SOCS-7, Nck, Ash and phospholipase C binding protein, Nck, Ash and phospholipase C gamma-binding protein, NCKAP4, Nck-associated protein 4, SOCS6, suppressor of cytokine signaling 7
Gene Symbols SOCS7
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SOCS7(suppressor of cytokine signaling 7) The peptide sequence was selected from the N terminal of SOCS7. Peptide sequence LDPKALPPGLALERTWGPAAGLEAQLAALGLGQPAGPGVKTVGGGCCPCP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30837
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.