Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOCS2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5119445

 View more versions of this product

Catalog No. PIPA5119445


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

SOCS2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the STAT-induced STAT inhibitor (SSI), also known as suppressor of cytokine signaling (SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including erythropoietin, GM-CSF, IL10 and interferon (IFN)-gamma. The protein encoded by this gene is found to interact with the cytoplasmic domain of insulin-like growth factor-1 receptor (IGF1R), and thus is thought to be involved in the regulation of IGF1R mediated cell signaling. Knockout studies in mice also suggested a regulatory role of this gene in IGF-1 related growth control.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOCS2
Polyclonal
Unconjugated
SOCS2
8030460M17; AI527257; AW108012; CIS2; CIS-2; Cish2; cytokine inducible SH2-containing protein 2; cytokine-inducible SH2 protein 2; D130043N08Rik; hg; JAB; Socs2; SOCS-2; SSI2; SSI-2; STAT induced STAT inhibitor-2; STATI2; STAT-induced STAT inhibitor 2; STAT-induced STAT inhibitor-2; Suppressor of cytokine signaling 2; suppressor of cytokine signaling-2
Rabbit
Antigen affinity chromatography, Protein A
RUO
100037966, 84607, 8835
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
O14508, O88582, Q7YRV6
SOCS2
Recombinant protein Suppressors Of Cytokine Signaling 2. The antigen corresponds to amino acid range 1-198 of the target protein.
200 μL
Primary
Human, Rat, Pig
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.