Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOCS2 Polyclonal Antibody

Rabbit Polyclonal Antibody

$237.00 - $539.50

Specifications

Antigen SOCS2
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119445
View Documents
Invitrogen™
PA5119445
200 μL
Each for $539.50
Only null left
Add to Cart
 
PIPA5121588
View Documents
Invitrogen™
PA5121588
20 μL
Each for $237.00
Only null left
Add to Cart
 
Description

Description

SOCS2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the STAT-induced STAT inhibitor (SSI), also known as suppressor of cytokine signaling (SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including erythropoietin, GM-CSF, IL10 and interferon (IFN)-gamma. The protein encoded by this gene is found to interact with the cytoplasmic domain of insulin-like growth factor-1 receptor (IGF1R), and thus is thought to be involved in the regulation of IGF1R mediated cell signaling. Knockout studies in mice also suggested a regulatory role of this gene in IGF-1 related growth control.
Specifications

Specifications

SOCS2
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Rat, Pig
SOCS2
8030460M17; AI527257; AW108012; CIS2; CIS-2; Cish2; cytokine inducible SH2-containing protein 2; cytokine-inducible SH2 protein 2; D130043N08Rik; hg; JAB; Socs2; SOCS-2; SSI2; SSI-2; STAT induced STAT inhibitor-2; STATI2; STAT-induced STAT inhibitor 2; STAT-induced STAT inhibitor-2; Suppressor of cytokine signaling 2; suppressor of cytokine signaling-2
SOCS2
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
O14508, O88582, Q7YRV6
100037966, 84607, 8835
Recombinant protein Suppressors Of Cytokine Signaling 2. The antigen corresponds to amino acid range 1-198 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.