Learn More
Description
Specifications
Specifications
| Antigen | SOD1/Cu-Zn SOD |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 101 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD1/Cu-Zn SOD (NP_000445.1; Ala 2-Gln 154). |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
