Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1/Cu-Zn SOD Antibody (3E5), Novus Biologicals™
SDP

Catalog No. NB298358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298358 50 μL
NB298359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298358 Supplier Novus Biologicals Supplier No. NBP32630950UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOD1/Cu-Zn SOD Monoclonal antibody specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen SOD1/Cu-Zn SOD
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 3E5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SOD1/Cu-Zn SOD [UniProt P00441]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6647
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.