Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1/Cu-Zn SOD Antibody, Novus Biologicals™
SDP

Catalog No. NB10060944 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060944 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060944 Supplier Novus Biologicals Supplier No. NB10060944
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

SOD1/Cu-Zn SOD Polyclonal specifically detects SOD1/Cu-Zn SOD in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.

Specifications

Antigen SOD1/Cu-Zn SOD
Applications Immunocytochemistry, Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01 - 0.03 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P00441
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Gene Symbols SOD1
Host Species Goat
Immunogen Peptide with sequence C-SRKHGGPKDEERH corresponding to internal region according to NP_000445.1.
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6647
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted cross-reactivity based on sequence identity: Dog.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.