Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1/Cu-Zn SOD Rabbit anti-Human, Clone: 106, Novus Biologicals™
SDP

Catalog No. NB021402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021402 50 μg
NB021401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021402 Supplier Novus Biologicals Supplier No. NBP28996950UL

Rabbit Monoclonal Antibody

SOD1/Cu-Zn SOD Monoclonal specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SOD1/Cu-Zn SOD
Applications Flow Cytometry
Classification Monoclonal
Clone 106
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Host Species Rabbit
Immunogen Recombinant Human SOD1/Cu-Zn SOD Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6647
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.