Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2/Mn-SOD Antibody (001), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB316851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316851 Supplier Novus Biologicals Supplier No. NBP290138MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen SOD2/Mn-SOD
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 001
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD2/Mn-SOD (Uniprot#: P04179-1; Lys 25-Lys 222).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6648
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.