Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2/Mn-SOD Antibody, Novus Biologicals™
SDP

Catalog No. NB1001969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001969 Supplier Novus Biologicals Supplier No. NB1001969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

SOD2/Mn-SOD Polyclonal specifically detects SOD2/Mn-SOD in Bovine, Canine, Chicken, Gerbil, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SOD2/Mn-SOD
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100
Formulation PBS (pH 7.4), 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P04179
Gene Alias EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial
Gene Symbols SOD2
Host Species Rabbit
Immunogen Recombinant Human Mn SOD Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6648
Test Specificity Detects approx 25 kDa.
Target Species Bovine, Canine, Chicken, Gerbil, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.