Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD3/EC-SOD Antibody (4GG11G6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP122417IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122417IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122417IR Supplier Novus Biologicals Supplier No. NBP122417IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOD3/EC-SOD Monoclonal specifically detects SOD3/EC-SOD in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SOD3/EC-SOD
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4GG11G6
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. P08294
Gene Alias EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular
Gene Symbols SOD3
Host Species Mouse
Immunogen Human extracellular SOD purified from aortas
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6649
Test Specificity Detects extracellular SOD approx 35kDa.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.