Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SODD/BAG4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056696 0.1 mg
NB10056696T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056696 Supplier Novus Biologicals Supplier No. NB10056696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

SODD/BAG4 Polyclonal specifically detects SODD/BAG4 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), KnockDown, Western Blot.

Specifications

Antigen SODD/BAG4
Applications Flow Cytometry, Flow Cytometry (Cell Surface), KnockDown, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Flow Cytometry 1:10-1:1000, Flow (Cell Surface) reported in scientific literature (PMID 15592361), KnockDown Validated
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. O95429
Gene Alias BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4
Gene Symbols BAG4
Host Species Rabbit
Immunogen This polyclonal antibody was raised against a peptide corresponding to amino acids 443-457 of human SODD protein.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitochondrial Mediated Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9530
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.