Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SODD/BAG4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$181.90 - $402.00
Specifications
Antigen | SODD/BAG4 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4 ug/ml, Flow Cytometry 1:10-1:1000, Flow (Cell Surface) reported in scientific literature (PMID 15592361), KnockDown Validated |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056696T
![]() |
Novus Biologicals
NB10056696SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056696
![]() |
Novus Biologicals
NB10056696 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
SODD/BAG4 Polyclonal specifically detects SODD/BAG4 in Human samples. It is validated for Western Blot, Flow Cytometry, Flow (Cell Surface), Knockdown Validated.Specifications
SODD/BAG4 | |
Western Blot 2-4 ug/ml, Flow Cytometry 1:10-1:1000, Flow (Cell Surface) reported in scientific literature (PMID 15592361), KnockDown Validated | |
Polyclonal | |
Purified | |
RUO | |
Human | |
O95429 | |
9530 | |
This polyclonal antibody was raised against a peptide corresponding to amino acids 443-457 of human SODD protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Mitochondrial Mediated Pathway, Tumor Suppressors | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4 | |
BAG4 | |
IgG | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title