Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%

Catalog No. AAA1091714
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 22767-50-6 | C7H15NaO3S | 202.244 g/mol

HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number MFCD00007543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
Beilstein 3731762
Solubility Information Soluble in water.
Formula Weight 202.24
Percent Purity 99%
Assay (dry wt.)
Chemical Name or Material Sodium 1-heptanesulfonate, Water <2%
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-210-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.