Learn More
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals
Description
Sodium 1-octanesulfonate monohydrate is used as an anionic detergent. It is an ion pairing reagent used in the HPLC analysis of proteins and peptides. Further, it is used as a surface active agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Quantity | 10 g |
Solubility Information | Soluble in water. |
Formula Weight | 234.29 (216.28 Anhydrous) |
Percent Purity | ≥99% |
Physical Form | Crystalline |
Chemical Name or Material | Sodium 1-octanesulfonate monohydrate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 226-195-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.