Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Propanesulfonate 98.0+%, TCI America™
SDP

Catalog No. I034125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 14533-63-2
Molecular Formula C3H7NaO3S
Molecular Weight (g/mol) 146.136
MDL Number MFCD00062546
InChI Key NPAWNPCNZAPTKA-UHFFFAOYSA-M
Synonym sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt
PubChem CID 4319363
IUPAC Name sodium;propane-1-sulfonate
SMILES CCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 255°C
Color White
Quantity 25 g
Formula Weight 146.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Propanesulfonate [Reagent for Ion-Pair Chromatography]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.