Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Tridecanesulfonate 98.0+%, TCI America™
SDP

Catalog No. I035125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5802-89-1
Molecular Formula C13H27NaO3S
Molecular Weight (g/mol) 286.406
MDL Number MFCD00024993
InChI Key CACJZDMMUHMEBN-UHFFFAOYSA-M
Synonym IPC-ALKS-13, 1-Tridecanesulfonic Acid Sodium Salt
PubChem CID 23683718
IUPAC Name sodium;tridecane-1-sulfonate
SMILES CCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 286.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Tridecanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-1640
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.