Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-nitrobenzenesulfonate, 98%, water <2%

Catalog No. AAA1024422
Click to view available options
Quantity:
100 g
500 g
2500 g

Catalog No. AAA1024422

Supplier: Thermo Scientific Chemicals A1024422

Only null left
Add to Cart

CAS: 127-68-4 | C6H4NNaO5S | 225.15 g/mol

Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

Solubility
Soluble in water 200 g/L (20C).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-68-4
Molecular Formula C6H4NNaO5S
Molecular Weight (g/mol) 225.15
MDL Number MFCD00007490
InChI Key LJRGBERXYNQPJI-UHFFFAOYSA-M
Synonym sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
PubChem CID 31389
IUPAC Name sodium;3-nitrobenzenesulfonate
SMILES C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

Melting Point 350°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water 200g/L (20C).
Formula Weight 225.16 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium 3-nitrobenzenesulfonate, Water <2%
Hazard Category H317-H319
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
EINECSNumber 204-857-3
RTECSNumber DB7195000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.