Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 5-Formyl-2-furansulfonate 98.0+%, TCI America™
SDP

Catalog No. F02685G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Catalog No. F02685G

Supplier: TCI America F02685G

Chemical Identifiers

CAS 31795-44-5
Molecular Formula C5H3NaO5S
Molecular Weight (g/mol) 198.12
MDL Number MFCD00003231
InChI Key ZTJAVDOZUQPVAV-UHFFFAOYSA-M
Synonym 5-Formyl-2-furansulfonic Acid Sodium Salt, Furfural-5-sulfonic Acid Sodium Salt
PubChem CID 3772036
IUPAC Name sodium 5-formylfuran-2-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(O1)C=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 198.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 5-Formyl-2-furansulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.